MAT333118861 Matrix Scientific

1-Ethyladamantane

  • MDL number MFCD00142626
  • Cas Number : 770-69-4
  • Molecular Weight : 164.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063682.html

Safety Information

MSDS http://www.matrixscientific.com/063682.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart