MAT333118867 Matrix Scientific

1-(1-Adamantyl)pyrrolidin-2-one

  • MDL number MFCD01749832
  • Cas Number : 3716-62-9
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063688.html

Safety Information

MSDS http://www.matrixscientific.com/063688.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart