MAT333118891 Matrix Scientific

4,5-Diiodo-1H-1,2,3-triazole

  • MDL number MFCD15146495
  • Cas Number : 28194-09-4
  • Molecular Weight : 320.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/063712.html

Safety Information

MSDS http://www.matrixscientific.com/063712.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart