MAT333119081 Matrix Scientific

Methyl 3-iodo-4-phenoxybenzoate

  • MDL number MFCD02912489
  • Cas Number : 100725-29-9
  • Molecular Weight : 354.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063902.html

Safety Information

MSDS http://www.matrixscientific.com/063902.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart