MAT333119097 Matrix Scientific

(3,5-Dimethylphenyl)acetic acid

  • MDL number MFCD00082776
  • Cas Number : 42288-46-0
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/063918.html

Safety Information

MSDS http://www.matrixscientific.com/063918.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart