MAT333119099 Matrix Scientific

(4,5-Dichloro-3-methyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD16618444
  • Cas Number : 1228551-78-7
  • Molecular Weight : 209.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063920.html

Safety Information

MSDS http://www.matrixscientific.com/063920.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart