MAT333119122 Matrix Scientific

Ethyl 6-methoxy-1H-indole-2-carboxylate

  • MDL number MFCD01544121
  • Cas Number : 15050-04-1
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063943.html

Safety Information

MSDS http://www.matrixscientific.com/063943.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart