MAT333119170 Matrix Scientific

2-[2-(1H-Pyrazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione

  • MDL number MFCD16618465
  • Cas Number : 1195687-62-7
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/063992.html

Safety Information

MSDS http://www.matrixscientific.com/063992.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart