MAT333119230 Matrix Scientific

3-Bromo-4-(difluoromethyl)pyridine

  • MDL number MFCD18257232
  • Cas Number : 114468-05-2
  • Molecular Weight : 208.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/064052.html

Safety Information

MSDS http://www.matrixscientific.com/064052.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart