MAT333119253 Matrix Scientific

4-Chloro-6-methoxypyridine-3-carboxylic acid

  • MDL number MFCD13188727
  • Cas Number : 1060806-60-1
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-224
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/064075.html

Safety Information

MSDS http://www.matrixscientific.com/064075.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart