MAT333119362 Matrix Scientific

Methyl 5,7-dimethyl-1H-indole-2-carboxylate

  • MDL number MFCD15201241
  • Cas Number : 1158249-28-5
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064184.html

Safety Information

MSDS http://www.matrixscientific.com/064184.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart