MAT333119417 Matrix Scientific

3-(5-Formyl-2-furyl)benzoic acid

  • MDL number MFCD00454916
  • Cas Number : 304884-54-6
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064239.html

Safety Information

MSDS http://www.matrixscientific.com/064239.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart