MAT333119422 Matrix Scientific

7-Acetyl-3,4-dihydroquinoxalin-2(1H)-one

  • MDL number MFCD17078769
  • Cas Number : 1255147-27-3
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064244.html

Safety Information

MSDS http://www.matrixscientific.com/064244.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart