MAT333119430 Matrix Scientific

(2-Amino-6-chloropyrimidin-4-yl)cyanamide

  • MDL number MFCD17267867
  • Cas Number : 1255147-20-6
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064252.html

Safety Information

MSDS http://www.matrixscientific.com/064252.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart