MAT333119441 Matrix Scientific

2-Chloro-N-[3-(trifluoromethyl)phenyl]pyrimidin-4-amine

  • MDL number MFCD11505045
  • Cas Number : 879127-23-8
  • Molecular Weight : 273.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064263.html

Safety Information

MSDS http://www.matrixscientific.com/064263.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart