MAT333119475 Matrix Scientific

Ethyl oxo(pyrrolidin-1-yl)acetate

  • MDL number MFCD02859932
  • Cas Number : 41600-22-0
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064297.html

Safety Information

MSDS http://www.matrixscientific.com/064297.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart