MAT333119590 Matrix Scientific

1-Cyclohexyloctahydropyrrolo[1,2-a]pyrazine

  • MDL number MFCD00449927
  • Cas Number : 155206-41-0
  • Molecular Weight : 208.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/064412.html

Safety Information

MSDS http://www.matrixscientific.com/064412.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart