MAT333119605 Matrix Scientific

3-Methyl-1H-1,2,4-triazole-5-thiol

  • MDL number MFCD00137841
  • Cas Number : 7271-44-5
  • Molecular Weight : 115.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064427.html

Safety Information

MSDS http://www.matrixscientific.com/064427.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart