MAT333119658 Matrix Scientific

Pyridine-2,6-diyldi(methylene)-bis(methylcarbamate)

  • MDL number MFCD00473700
  • Cas Number : 1882-26-4
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/064480.html

Safety Information

MSDS http://www.matrixscientific.com/064480.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart