MAT333119659 Matrix Scientific

3,3',5,5'-Tetrabromobiphenyl-2,2',4,4'-tetrol

  • MDL number MFCD00427055
  • Cas Number : 27951-69-5
  • Molecular Weight : 533.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 277-278
Country of Origin USA
Product Link http://www.matrixscientific.com/064481.html

Safety Information

MSDS http://www.matrixscientific.com/064481.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart