MAT333119676 Matrix Scientific

2-Methoxyethyl 4-(3-hydroxyphenyl)-7-(2-methoxyphenyl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline

  • MDL number MFCD03312466
  • Cas Number : 599150-20-6
  • Molecular Weight : 463.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064498.html

Safety Information

MSDS http://www.matrixscientific.com/064498.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart