MAT333119762 Matrix Scientific

N-(2-Carboxyethyl)-N-methyl-beta-alanine

  • MDL number MFCD16301579
  • Cas Number : 21555-95-3
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156
Country of Origin USA
Boiling point 137-140� Torr
Product Link http://www.matrixscientific.com/064584.html

Safety Information

MSDS http://www.matrixscientific.com/064584.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart