MAT333119782 Matrix Scientific

Diethyl 4,4-dicyanoheptanedioate

  • MDL number MFCD18064638
  • Cas Number : 29668-96-0
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/064604.html

Safety Information

MSDS http://www.matrixscientific.com/064604.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart