MAT333119792 Matrix Scientific

1-(Trifluoroacetyl)-2,5-dihydro-1H-pyrrole

  • MDL number MFCD18064643
  • Cas Number : 111185-41-2
  • Molecular Weight : 165.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064614.html

Safety Information

MSDS http://www.matrixscientific.com/064614.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart