MAT333119839 Matrix Scientific

4-(Pyrrolidin-1-ylmethyl)benzaldehyde

  • MDL number MFCD07782421
  • Cas Number : 650628-72-1
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064662.html

Safety Information

MSDS http://www.matrixscientific.com/064662.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart