MAT333119888 Matrix Scientific

5-(4-tert-Butylphenyl)cyclohexane-1,3-dione

  • MDL number MFCD13873932
  • Cas Number : 946686-29-9
  • Molecular Weight : 244.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064711.html

Safety Information

MSDS http://www.matrixscientific.com/064711.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart