MAT333119946 Matrix Scientific

(7,8-Dihydroxy-4-methyl-2-oxo-2H-chromen-3-yl)-acetic acid

  • MDL number MFCD03788880
  • Cas Number : 773866-17-4
  • Molecular Weight : 250.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 277
Country of Origin USA
Product Link http://www.matrixscientific.com/064769.html

Safety Information

MSDS http://www.matrixscientific.com/064769.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart