MAT333119947 Matrix Scientific

Methyl (5,7-dihydroxy-4-methyl-2-oxo-2H-chromen-3-yl)acetate

  • MDL number MFCD08741832
  • Cas Number : 892559-73-8
  • Molecular Weight : 264.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/064770.html

Safety Information

MSDS http://www.matrixscientific.com/064770.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart