MAT333119949 Matrix Scientific

4-(4-Chlorophenyl)pyrimidin-2-amine

  • MDL number MFCD00665913
  • Cas Number : 133256-51-6
  • Molecular Weight : 205.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064772.html

Safety Information

MSDS http://www.matrixscientific.com/064772.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart