MAT333120049 Matrix Scientific

1-Benzyl-1H-indole-5-carboxylic acid

  • MDL number MFCD11108892
  • Cas Number : 1030423-92-7
  • Molecular Weight : 251.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064872.html

Safety Information

MSDS http://www.matrixscientific.com/064872.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart