MAT333120074 Matrix Scientific

(2S)-2-(3-Oxo-1,2-benzisothiazol-2(3H)-yl)-3-phenylpropanoic acid

  • MDL number MFCD11868499
  • Cas Number : 1212403-91-2
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064897.html

Safety Information

MSDS http://www.matrixscientific.com/064897.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart