MAT333120092 Matrix Scientific

7-Chloro-1,3-dihydroxy-9H-xanthen-9-one

  • MDL number MFCD11108884
  • Cas Number : 100334-95-0
  • Molecular Weight : 262.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/064915.html

Safety Information

MSDS http://www.matrixscientific.com/064915.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart