MAT333120096 Matrix Scientific

1,3-Dihydroxy-5-methyl-9H-xanthen-9-one

  • MDL number MFCD06766389
  • Cas Number : 50493-39-5
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280
Country of Origin USA
Product Link http://www.matrixscientific.com/064919.html

Safety Information

MSDS http://www.matrixscientific.com/064919.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart