MAT333120274 Matrix Scientific

3-Ethyl-7-hydroxy-4-methyl-2H-chromen-2-one

  • MDL number MFCD00449861
  • Cas Number : 53666-71-0
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065099.html

Safety Information

MSDS http://www.matrixscientific.com/065099.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart