MAT333120517 Matrix Scientific

Methyl N-(trifluoroacetyl)tryptophanate

  • MDL number MFCD19103282
  • Cas Number : 1604-49-5
  • Molecular Weight : 314.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/065348.html

Safety Information

MSDS http://www.matrixscientific.com/065348.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart