MAT333120522 Matrix Scientific

1,4-Dimethyl-9H-carbazole-3-carbaldehyde

  • MDL number MFCD00189389
  • Cas Number : 14501-66-7
  • Molecular Weight : 223.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/065353.html

Safety Information

MSDS http://www.matrixscientific.com/065353.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart