MAT333120557 Matrix Scientific

2,5-Diphenylthiophene

  • MDL number MFCD00626285
  • Cas Number : 1445-78-9
  • Molecular Weight : 236.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065388.html

Safety Information

MSDS http://www.matrixscientific.com/065388.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart