MAT333120585 Matrix Scientific

4-[(Methylamino)methyl]benzoic acid

  • MDL number MFCD08756336
  • Cas Number : 96084-38-7
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/065416.html

Safety Information

MSDS http://www.matrixscientific.com/065416.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart