MAT333120682 Matrix Scientific

1-Methyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD00460857
  • Cas Number : 98711-43-4
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065513.html

Safety Information

MSDS http://www.matrixscientific.com/065513.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart