MAT333120714 Matrix Scientific

Ethyl 4-(3,4-dimethoxyphenyl)-2,4-dioxobutanoate

  • MDL number MFCD03411532
  • Cas Number : 70909-46-5
  • Molecular Weight : 280.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065545.html

Safety Information

MSDS http://www.matrixscientific.com/065545.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart