MAT333120752 Matrix Scientific

4-Bromo-2-isopropyl-1H-imidazole

  • MDL number MFCD19103367
  • Cas Number : 1256643-65-8
  • Molecular Weight : 189.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065583.html

Safety Information

MSDS http://www.matrixscientific.com/065583.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart