MAT333120863 Matrix Scientific

1-Isopropyl-5-oxopyrrolidine-3-carbonyl chloride

  • MDL number MFCD11939071
  • Cas Number : 1181455-29-7
  • Molecular Weight : 189.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/065695.html

Safety Information

MSDS http://www.matrixscientific.com/065695.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart