MAT333120946 Matrix Scientific

1-tert-Butyl 2-methyl 2-methylpyrrolidine-1,2-dicarboxylate

  • MDL number MFCD18642785
  • Cas Number : 317355-80-9
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065779.html

Safety Information

MSDS http://www.matrixscientific.com/065779.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart