MAT333121040 Matrix Scientific

(4,4,6,8-Tetramethyl-2-oxo-5,6-dihydro-4H-pyrrolo-[3,2,1-ij]quinolin-1(2H)-ylidene)malononitrile

  • MDL number MFCD03903957
  • Cas Number : 674363-06-5
  • Molecular Weight : 291.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065874.html

Safety Information

MSDS http://www.matrixscientific.com/065874.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart