MAT333121124 Matrix Scientific

5-(3-Methylphenyl)cyclohexane-1,3-dione

  • MDL number MFCD08753427
  • Cas Number : 762243-26-5
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065962.html

Safety Information

MSDS http://www.matrixscientific.com/065962.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart