MAT333121149 Matrix Scientific

6-Bromo-3-(4-chlorophenyl)quinazoline-2,4(1H,3H)-dione

  • MDL number MFCD00262935
  • Cas Number : 23965-02-8
  • Molecular Weight : 351.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/065987.html

Safety Information

MSDS http://www.matrixscientific.com/065987.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart