MAT333121181 Matrix Scientific

N-(2,4-Dichlorophenyl)-2-mercaptoacetamide

  • MDL number MFCD00847296
  • Cas Number : 60466-54-8
  • Molecular Weight : 236.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066019.html

Safety Information

MSDS http://www.matrixscientific.com/066019.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart