MAT333121259 Matrix Scientific

N-(2-Chlorophenyl)-N'-(6-methyl-4-oxo-1,4-dihydropyrimidin-2-yl)guanidine

  • MDL number MFCD19103553
  • Cas Number : 857413-65-1
  • Molecular Weight : 277.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/066097.html

Safety Information

MSDS http://www.matrixscientific.com/066097.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart