MAT333121377 Matrix Scientific

4,5,6,7-Tetrahydro-2H-indazole

  • MDL number MFCD00462153
  • Cas Number : 2305-79-5
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066216.html

Safety Information

MSDS http://www.matrixscientific.com/066216.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart