MAT333121527 Matrix Scientific

6-Methyl-2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carboxylic acid

  • MDL number MFCD16070480
  • Cas Number : 1249049-37-3
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/066368.html

Safety Information

MSDS http://www.matrixscientific.com/066368.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart